missing translation for 'onlineSavingsMsg'
Learn More

Pectin, MP Biomedicals™

Product Code. 11488210 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488210 500 g 500g
11478210 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11488210 Supplier MP Biomedicals Supplier No. 0210258790
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 9000-69-5
Molecular Formula (C7H10O7)n
Synonym Poly-D-galacturonic acid methyl ester

Specifications

Color Beige to White
Physical Form Powder
Quantity 500 g
Formula Weight 20,000 to 400,000
Chemical Name or Material Pectin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.