missing translation for 'onlineSavingsMsg'
Learn More

Pectin (Technical Grade), TRC

Product Code. 30424546
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30424546 100 g
30416144 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30424546 Supplier Toronto Research Chemicals Supplier No. TRCP218305100G
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9000-69-5
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
Synonym AF 701, AP 40, AS 511, Amid CS 005, BIG-J, Beta BI-J, Beta-Pectin, Cesapectin, Classic AF 501, Classic AM 201, Classic Cu 701, Colyer Pectin, D 225, D-D Slowset, E 440, E 440a, Fluka 7682, GelSite, Genu 8003, Genu Beta Pectin, Genu JMJ 100, Genu Pectin, Genu Pectin 121J, Genu Pectin 121J Slow Set, Genu Pectin A, Genu Pectin BIG-J, Genu Pectin DD Slow Set J, Genu Pectin DSS, Genu Pectin L 200, Genu Pectin LM 101AS, Genu Pectin LM 104AS, Genu Pectin LM 104AS-FS, Genu Pectin LM 104AS-J, Genu Pectin LM 105AS, Genu Pectin LM 105S, Genu Pectin LM 18, Genu Pectin LM 18CG-Z, Genu Pectin LM 85AS, Genu Pectin USP-H, Genu Pectin X 0905, Genu Pectin YM 100, Genu Pectin YM 150J, Genu pHresh DF, Genu Pectin HM-USP-H, Genu Pectin LM 12CG, Genu Pectin LM5 CS, Genue LM 12CG, H&F Pectin Classic AF 701, H&F Pectin Classic CU 701, HMP RS400, Herbacel Citrus Fiber, JMJ-J, LM 101AS-J, LM 102AS, LM 104AS-FS, LM 104AS-J, LM 12CG-Z, LMNA/P 3450NA95, LNSN 325, MCP, Marpee NL, Marpee OM, Methoxypectin, Methyl Pectin, Methyl Pectinate, MexPec 1200, MexPec 1400, MexPec 1500, Mexpectin XSS 100, NCB-PE 40, Neosoft P, Newgelin LP 4, OB 700, OF 305, OXPEC, PE 2006MB1, PG 717CS, Pectin 1694, Pectin Classic CU 701, Pectin D 210, Pectin JM 150JN, Pectinate, Pectinic Acid, Pectins, Pecto, Pepidol PEG, Pomelite Lv, Pomosin LM 12, Pomosin LM 12CG-Z, Red Ribbon 3G, SM 478, Sango-stop, Slendid 120, Slendid 200, Slendid L 200, Splendid, TIC 1200, TIC Pretested Pectin 1460 Powder, TIC Pretested Pre-hydrated 1694 Powder, USP 100, Unipectin, Unipectin AYD 30, Unipectin OB 70, Unipectine 150RS150C, Unipectine 3450NA95, Unipectine 903, Unipectine AMP 285, Unipectine AYD 30T, Unipectine AYD 358, Unipectine LM OF 300C, Unipectine MRS 150, Unipectine OF 463, Unipectine RS 150° SAG, Unipectine RS ND, Unipectine SS 150, Unipectine SS-ND, VIS TOP D 2122, VIS TOP D 2250, X 939-04X, YM 115H, YM 115LJ, YM 15LJ
SMILES O[C@H]([C@H]1O)[C@@H](OC)[C@@H](C(O)=O)O[C@@H]1O[C@H]([C@@H]([C@H]2O)O)[C@@H](C(O)=O)O[C@@H]2O[C@H]([C@@H]([C@H]3O)O)[C@@H](C(O)=O)O[C@@H]3O[C@H]([C@@H]([C@H]4O)O)[C@@H](C(O)=O)O[C@@H]4OC

Specifications

Quantity 100 g
InChI Formula InChI=1S/C5H10O5/c6-1-3(8)5(10)4(9)2-7/h1,3-5,7-10H,2H2
Chemical Name or Material Pectin (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.