Learn More
PEG (methoxy group) Rabbit, Clone: RM105, Abnova™
Rabbit monoclonal antibody raised against polyethylene glycol (methoxy group).
Brand: Abnova MAB12825
Additional Details : Weight : 0.00010kg
Specifications
PEG (methoxy group) | |
Monoclonal | |
Unconjugated | |
ELISA (0.01-0.3 ug/mL) Flow Cytometry Immunocytochemistry Immunohistochemistry (0.5-2 ug/mL) Immunoprecipitation Western Blot (0.05-1 ug/mL) The optimal working dilution should be determined by the end user. | |
Rabbit | |
Protein A affinity purification | |
RUO | |
This antibody reacts to the methoxy group of Polyethylene glycol (PEG). It is specific for methoxypoly (ethylene glycol). | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
RM105 | |
Rabbit monoclonal antibody raised against polyethylene glycol (methoxy group). | |
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
Polyethylene glycol (methoxy group) conjugated with KLH. | |
100 μg | |
Primary | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Safety and Handling
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P233-Keep container tightly closed.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P404-Store in a closed container.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium phosphate monobasic, potassium phosphate dibasic