Learn More
Pemetrexed Disodium Heptahydrate, Mikromol™
Description
Pemetrexed Disodium Heptahydrate
Specifications
Specifications
| Chemical Name or Material | Pemetrexed disodium heptahydrate |
| CAS | 357166-29-1 |
| Analyte or Component Names | Pemetrexed disodium heptahydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H19 N5 O6 . 2 Na . 7 H2 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | L-Glutamic acid, N-[4-[2-(2-amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-, sodium salt, hydrate (1:2:7), L-Glutamic acid, N-[4-[2-(2-amino-4,7-dihydro-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-, disodium salt, heptahydrate (9CI), Pemetrexed disodium heptahydrate, Disodium (2S)-2-[[4-[2-(2-amino-4-oxo-4,7-dihydro-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioate heptahydrate |
| InChI Formula | InChI=1S/C20H21N5O6.2Na.7H2O/c21-20-24-16-15(18(29)25-20)12(9-22-16)6-3-10-1-4-11(5-2-10)17(28)23-13(19(30)31)7-8-14(26)27;;;;;;;;;/h1-2,4-5,9,13H,3,6-8H2,(H,23,28)(H,26,27)(H,30,31)(H4,21,22,24,25,29);;;7*1H2/q;2*+1;;;;;;;/p-2/t13-;;;;;;;;;/m0........./s1 |
| SMILES | O.O.O.O.O.O.O.[Na+].[Na+].NC1=NC(=O)c2c(CCc3ccc(cc3)C(=O)N[C@@H](CCC(=O)[O-])C(=O)[O-])c[nH]c2N1 |
| IUPAC Name | disodium;(2S)-2-[[4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioate;heptahydrate |
| Show More |