missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Penciclovir

Product Code. 17211286 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17211286 1 g 1g
17221286 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 17211286 Supplier Thermo Scientific Chemicals Supplier No. 468730010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39809-25-1
Molecular Formula C10H15N5O3

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: 1.7 mg/mL (20°C).
Melting Point 275.0°C to 277.0°C
Formula Weight 253.26
InChI Key JNTOCHDNEULJHD-UHFFFAOYSA-N
SMILES NC1=NC(=O)C2=C(N1)N(CCC(CO)CO)C=N2
IUPAC Name 2-amino-9-[4-hydroxy-3-(hydroxymethyl)butyl]-6,9-dihydro-3H-purin-6-one
Molecular Weight (g/mol) 253.26
Chemical Name or Material Penciclovir

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.