missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Penicillin G-d5 Potassium Salt (phenyl-d5), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Penicillin G-d5 Potassium Salt (phenyl-d5) |
| Molecular Formula | C16 D5 H12 N2 O4 S . K |
| Quantity | 0.005 g |
| Synonym | Penicillin G Potassium D5 (Phenyl D5) |
| InChI Formula | InChI=1 S/C16H18N2O4S.K/c1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9;/h3-7,11-12,14 H,8H2,1-2H3,(H,17,19)(H,21,22);/q;+1/p-1/t11-,12+,14-;/m1./s1/i3D,4 D,5 D,6 D,7 D; |
| SMILES | [K+].[2 H]c1c([2 H])c([2 H])c(CC(=O)N[C@H]2 [C@H]3 SC(C)(C)[C@@H](N3C2=O)C(=O)[O-])c([2 H])c1[2 H] |
| IUPAC Name | potassium;(2 S,5 R,6 R)-3,3-dimethyl-7-oxo-6-[[2-(2,3,4,5,6-pentadeuteriophenyl)acetyl]amino]-4-thia-1-azabicyclo[3.2.0 ]heptane-2-carboxylate |
| Molecular Weight (g/mol) | 377.511 |
| Formula Weight | 377.086 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction