Learn More
Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, & nanoViper™ Fittings
Product Code. 17437583 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Cartridge Holder Kit
Tubing
Includes:
Cartridge Holder, Tubing, Fittings
Tubing, Fittings
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Includes Unit Size
17437583 Cartridge Holder Kit Cartridge Holder, Tubing, Fittings Each
17427583 Tubing Tubing, Fittings Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17437583 Supplier Thermo Scientific™ Supplier No. 174502
Big Savings for Better Separation
Use promo code 27992
View all promotions
Estimated Shipment: 05-11-2026
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, and nanoViper™ Fittings to connect your PepMap™ Neo Trap Cartridge to your HPLC system.
Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, and nanoViper™ Fittings to connect your PepMap Neo Trap Cartridge (Catalog Number 174500) to your HPLC system.
See Viper and nanoViper resources
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 30 μm (tubing)
For Use With PepMap™ Neo Trap Cartridge (part number 174500)
Includes Cartridge Holder, Tubing, Fittings
Length (Metric) 5 mm
Max. Pressure 22,000 psi (1500 bar)
Quantity Each
Temperature 60°C
Product Line PepMap Neo
Description Nano Trap Cartridge Holder with PEEK Tubing and nanoViper™ Fittings
For Use With (Application) Trap and Elute, Peptide Mapping
Product Type Cartridge Holder Kit