Learn More
Invitrogen™ Pepsinogen I Recombinant Rabbit Monoclonal Antibody (6L10S1)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558064
Description
Immunogen sequence: MKWLLLLGLV ALSECIMYKV PLIRKKSLRR TLSERGLLKD FLKKHNLNPA RKYFPQWEAP TLVDEQPLEN YLDMEYFGTI GIGTPAQDFT VVFDTGSSNL WVPSVYCSSL ACTNHNRFNP EDSSTYQSTS ETVSITYGTG SMTGILGYDT VQVGGISDTN QIFGLSETEP GSFLYYAPFD GILGLAYPSI SSSGATPVFD NIWNQGLVSQ DLFSVYLSAD DQSGSVVIFG GIDSSYYTGS LNWVPVTVEG YWQITVDSIT MNGEAIACAE GCQAIVDTGT SLLTGPTSPI ANIQSDIGAS ENSDGDMVVS CSAISSLPDI VFTINGVQYP VPPSAYILQS EGSCISGFQG MNLPTESGEL WILGDVFIRQ YFTVFDRANN QVGLAPVA.
PGA5: Pepsinogen 5, group I (pepsinogen A). Pepsinogens are the inactive precursors of pepsin, the major acid protease found in the stomach. Pepsin is one of the main proteolytic enzymes secreted by the gastric mucosa. Pepsin consists of a single polypeptide chain and arises from its precursor, pepsinogen, by removal of a 41 amino acid segment from the N-terminus. Pepsinogen is synthesized in the stomach lining, and hydrochloric acid, also produced by the gastric mucosa, is necessary to convert the inactive enzyme and to maintain the optimum acidity (pH 1-3) for pepsin function. Pepsin is particularly effective in cleaving peptide bonds involving aromatic amino acids. Pepsin shows extremely broad specificity; although bonds involving phenylalanine and leucine are preferred, many others are also cleaved to some extent. PGA5 is a member of the subfamily A1 within the pepsin family and is the predominant endopeptidase in the gastric juice of vertebrates. PGA5 is inhibited by ovUS-1, a uterine serpin.
Specifications
| Pepsinogen I | |
| Recombinant Monoclonal | |
| 1.01 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q9D106 | |
| Pga5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-388 of human Pepsinogen I (PG I) (NP_001073276.1). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 6L10S1 | |
| Unconjugated | |
| Pga5 | |
| 1110035E17Rik; Pepf; pepsin A; pepsin A-3; Pepsin A-5; Pepsin F; pepsinogen 3, group I (pepsinogen A); pepsinogen 5, group I; pepsinogen 5, group I (pepsinogen A); pepsinogen A3; pepsinogen A5; pepsinogen F; pepsinogen F protein; pepsinogen-3; Pepsinogen-5; Pg5; PGA3; PGA5 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 58803, 60372 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.