Learn More
Honeywell™ PERFECT POLY™ PU First Gray Gloves
Description
- Gray polyamide knitted glove
- Gray polyurethane coating at the palm and fingertips
- The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
- The glove offers maximum comfort and added sturdiness
- The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The ventilated back and PU coating optimise the glove's breathability
- Polyamide with its polyurethane coating provide excellent abrasion and tear resistance
- The PU coating ensures a good grip
- Non pilling, the polyamide is suitable for precision work
- The gloves are certified as silicone-free to ensure their use in painting environment
- Gray color makes it more soilproof, thus ensuring a longer life
Specifications
Specifications
| Certifications/Compliance | EN 420-2003 Basic standard, EN 388-2016 Mechanical risk |
| Description | Knitted Coating, 13 gauge |
| Coating Material | Polyurethane |
| Color | Gray |
| Material | Polyamide |
| Size | 9 |
| Length (Metric) | 25 cm |