missing translation for 'onlineSavingsMsg'
Learn More

Perfluoroheptanesulfonic Acid Sodium Salt, TRC

Product Code. 30007646
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007646 5 mg 5mg
30135422 50 mg 50mg
30136690 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007646 Supplier Toronto Research Chemicals Supplier No. TRCP6981965MG
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21934-50-9
Molecular Formula C7 F15 O3 S . Na
Molecular Weight (g/mol) 472.1
Synonym Perfluoroheptane Sulfonic Acid Sodium, 1-Heptanesulfonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoro-, sodium salt (8CI,9CI), Sodium perfluoroheptanesulfonate, 1-Heptanesulfonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoro-, sodium salt (8CI,9CI), Sodium perfluoroheptanesulfonate, Perfluoroheptanesulfonate, sodium salt, PFHpS, sodium salt
IUPAC Name sodium;1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoroheptane-1-sulfonate
SMILES [Na+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C7HF15O3S.Na/c8-1(9,2(10,11)4(14,15)6(18,19)20)3(12,13)5(16,17)7(21,22)26(23,24)25;/h(H,23,24,25);/q;+1/p-1
Formula Weight 471.9226
Percent Purity >95%
Chemical Name or Material Perfluoroheptanesulfonic acid sodium salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.