missing translation for 'onlineSavingsMsg'
Learn More

Perfluorohexanesulfonic Acid (50μg/mL in Methanol) (Technical Grade), TRC

Product Code. 30465084
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30465084 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30465084 Supplier Toronto Research Chemicals Supplier No. TRCKIT81051X1ML
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 355-46-4
Molecular Formula C6 H F13 O3 S
Quantity 1 mL
Synonym 1-Hexanesulfonic acid, tridecafluoro- (6CI,8CI), 1,1,2,2,3,3,4,4,5,5,6,6,6-Tridecafluoro-1-hexanesulfonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,6-Tridecafluorohexane-1-sulfonic acid, 1-Perfluorohexanesulfonic acid, PFHxS
InChI Formula InChI=1S/C6HF13O3S/c7-1(8,3(11,12)5(15,16)17)2(9,10)4(13,14)6(18,19)23(20,21)22/h(H,20,21,22)
SMILES OS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane-1-sulfonic acid
Molecular Weight (g/mol) 400.11
Formula Weight 399.9439
Concentration or Composition (by Analyte or Components) Perfluorohexanesulfonic acid 50 μg/mL
Chemical Name or Material Perfluorohexanesulfonic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.