Learn More
B Braun™ Perfusor™ Extension Lines
Product Code. 10869620
Change view
Click to view available options
Length (Metric):
500 mm
1000 mm
Material:
Polyethylene
Volume (Metric):
0.4 mL (Priming)
0.8 mL (Priming)
Unit Size:
Each
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Material Volume (Metric) unitSize
10869620 1000 mm Polyethylene 0.8 mL (Priming) Each
10264934 500 mm Polyethylene 0.4 mL (Priming) Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10869620 Supplier B Braun™ Supplier No. 8255067
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
Constructed of Polyethylene or PVC. B Braun™ Extension Lines allow minimal dead space volume. Transparent tube provides accurate liquid visual.
  • Transparent tube provides accurate liquid visual
  • Minimal dead space volume
  • Luer-Lock connector
  • Can be used with syringe pumps up to a pressure of <2 bar
Specifications
Material Polyethylene
Color Transparent
Diameter (Metric) Inner Needle 1.0 mm
For Use With (Application) Mobilizing patients
Length (Metric) 1000 mm
Product Type IV Extension Line
Volume (Metric) 0.8 mL (Priming)