missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perilipin Rabbit anti-Mouse, Rat, Clone: 1K4W3, Novus Biologicals™
Rabbit Monoclonal Antibody
214.00€ - 520.00€
Specifications
| Antigen | Perilipin |
|---|---|
| Clone | 1K4W3 |
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18379641
|
Novus Biologicals
NBP3-16480-20UL |
20 μg |
214.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18325704
|
Novus Biologicals
NBP3-16480-100UL |
100 μg |
520.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Perilipin Monoclonal antibody specifically detects Perilipin in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Perilipin | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| PERI, perilipin, perilipin 1, perilipin-1, PLINLipid droplet-associated protein | |
| A synthetic peptide corresponding to a sequence within amino acids 400-500 of human Perilipin (O60240). HYVPLPRLSLMEPESEFRDIDNPPAEVERREAERRASGAPSAGPEPAPRLAQPRRSLRSAQSPGAPPGPGLEDEVATPAAPRPGFPAVPREKPKRRVSDSF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 1K4W3 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Diabetes Research, Lipid and Metabolism, Lipid Droplets | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 5346 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title