missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Periostin/OSF-2 Antibody (CL12087) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44358
This item is not returnable.
View return policy
Description
Periostin/OSF-2 Monoclonal antibody specifically detects Periostin/OSF-2 in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence,Immunohistochemistry
Specifications
| Periostin/OSF-2 | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| MGC119510, MGC119511, OSF-2osteoblast specific factor 2 (fasciclin I-like), OSF2periodontal ligament-specific periostin, Osteoblast-specific factor 2, PDLPOSTN, periostin, periostin isoform thy2, periostin isoform thy4, periostin isoform thy6, periostin isoform thy8, periostin, osteoblast specific factor, PNRP11-412K4.1 | |
| This antibody was generated using a synthetic peptide of Q15063, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Cardiovascular Biology, Extracellular Matrix | |
| 10631 | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence, Immunohistochemistry | |
| CL12087 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction