Learn More
Periostin/OSF-2 Antibody (CL12087) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-15128JF669
Product Code. 30296613
  • 737.00€ / 0.10mL
Description
Periostin/OSF-2 Monoclonal antibody specifically detects Periostin/OSF-2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Periostin/OSF-2
Monoclonal
Janelia Fluor 669
MGC119510, MGC119511, OSF-2osteoblast specific factor 2 (fasciclin I-like), OSF2periodontal ligament-specific periostin, Osteoblast-specific factor 2, PDLPOSTN, periostin, periostin isoform thy2, periostin isoform thy4, periostin isoform thy6, periostin isoform thy8, periostin, osteoblast specific factor, PNRP11-412K4.1
This antibody was developed using a synthetic peptide derived from Q15063, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cardiovascular Biology, Extracellular Matrix
10631
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL12087
50mM Sodium Borate
Mouse
Protein A purified