missing translation for 'onlineSavingsMsg'
Learn More

Permanent Yellow Rn, TRC

Product Code. 30179480
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179480 10 mg 10mg
30179247 100 mg 100mg
30179071 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179480 Supplier Toronto Research Chemicals Supplier No. TRCP54104010MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6528-34-3
Molecular Formula C18H18N4O6
Molecular Weight (g/mol) 386.36
IUPAC Name 2-[(E)-(4-methoxy-2-nitrophenyl)diazenyl]-N-(2-methoxyphenyl)-3-oxobutanamide
SMILES COc1ccc(N=NC(C(=O)C)C(=O)Nc2ccccc2OC)c(c1)[N+](=O)[O-]

Specifications

Quantity 10 mg
Synonym Butanamide, 2-[2-(4-methoxy-2-nitrophenyl)diazenyl]-N-(2-methoxyphenyl)-3-oxo-, Butanamide, 2-[(4-methoxy-2-nitrophenyl)azo]-N-(2-methoxyphenyl)-3-oxo- (9CI), 2-[2-(4-Methoxy-2-nitrophenyl)diazenyl]-N-(2-methoxyphenyl)-3-oxobutanamide, 2-[2-(4-Methoxy-2-nitrophenyl)diazen-1-yl]-N-(2-methoxyphenyl)-3-oxobutanamide, Acetanil Yellow RTCA, C.I. 11740, C.I. Pigment Yellow 65, Dalamar Yellow YT 820D, Dalamar Yellow YT-820-D, Hansa Yellow 3RN, Hansa Yellow RN, Permanent Yellow RN, Permansa Yellow Lightfast Medium RA 12187, Pigment Yellow 65, Sunbrite Yellow 65
InChI Formula InChI=1S/C18H18N4O6/c1-11(23)17(18(24)19-14-6-4-5-7-16(14)28-3)21-20-13-9-8-12(27-2)10-15(13)22(25)26/h4-10,17H,1-3H3,(H,19,24)
Formula Weight 386.1226
Chemical Name or Material Permanent Yellow Rn
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.