missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peroxiredoxin 4 Antibody (SAIC-40A-24), DyLight 755 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20105IR
This item is not returnable.
View return policy
Description
Peroxiredoxin 4 Monoclonal antibody specifically detects Peroxiredoxin 4 in Human samples. It is validated for Western Blot, ELISA
Specifications
| Peroxiredoxin 4 | |
| Monoclonal | |
| DyLight 755 | |
| Antioxidant enzyme AOE372, AOE37-2PRX-4, EC 1.11.1.15, peroxiredoxin 4, Peroxiredoxin IV, peroxiredoxin-4, prx4, prx-4, Prx-IV, thioredoxin peroxidase (antioxidant enzyme), Thioredoxin peroxidase AO372, Thioredoxin-dependent peroxide reductase A0372 | |
| Peptide "e;QITLNDLPVGR"e;, derived from the cell redox-regulator Peroxiredoxin-4, conjugated to KLH. | |
| 0.1 mL | |
| Signal Transduction | |
| 10549 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Western Blot, ELISA | |
| SAIC-40A-24 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction