missing translation for 'onlineSavingsMsg'
Learn More

Perphenazine, TRC

Product Code. 30135536
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135536 5 g 5g
30006328 1 g 1g
30011747 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135536 Supplier Toronto Research Chemicals Supplier No. TRCP2911005G
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-39-9
Molecular Formula C21 H26 Cl N3 O S
Molecular Weight (g/mol) 403.97
Synonym 1-Piperazineethanol, 4-[3-(2-chlorophenothiazin-10-yl)propyl]- (8CI), 4-[3-(2-Chloro-10H-phenothiazin-10-yl)propyl]-1-piperazineethanol, 1-(2-Hydroxyethyl)-4-[3-(2-chloro-10-phenothiazinyl)propyl]piperazine, 2-Chloro-10-3-[1-(2-hydroxyethyl)-4-piperazinyl] propyl phenothiazine, 2-Chloro-10-[3-[4-(2-hydroxyethyl)piperazin-1-yl]propyl]phenothiazine, Chloriprozine, Chlorperphenazine, Decentan, Emesinal, Etaperazin, Etaperazine, Ethaperazine, Fentazin, Fentazin syrup, Fentazine duolets, Mutabom, NSC 150866, PZC, PZC (tranquilizer), Perfenazine, Perfenil, Perphenan, Perphenazin, Perphenazine, Sch 3940, Thilatazin, Tranquisan, Trifaron, Trilafon, Trilifan, Triphenot, γ-[4-(β-Hydroxyethyl)piperazin-1-yl]propyl-2-chlorophenothiazine
IUPAC Name 2-[4-[3-(2-chlorophenothiazin-10-yl)propyl]piperazin-1-yl]ethanol
SMILES OCCN1CCN(CCCN2c3ccccc3Sc4ccc(Cl)cc24)CC1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C21H26ClN3OS/c22-17-6-7-21-19(16-17)25(18-4-1-2-5-20(18)27-21)9-3-8-23-10-12-24(13-11-23)14-15-26/h1-2,4-7,16,26H,3,8-15H2
Formula Weight 403.1485
Chemical Name or Material Perphenazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.