Learn More
PEX14 (Human Specific) Rabbit anti-Human, Unconjugated, Clone: 6K5, Proteintech
Rabbit Recombinant Antibody
149.00€ - 439.00€
Specifications
| Antigen | PEX14 (Human Specific) |
|---|---|
| Clone | 6K5 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17422453
|
Proteintech
80196-1-RR-20UL |
20 μL |
149.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17412453
|
Proteintech
80196-1-RR-100UL |
100 μL |
439.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PEX14 (human specific) antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human PEX14 (human specific). This antibody recognizes human antigen. The PEX14 (human specific) antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| PEX14 (Human Specific) | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Human | |
| IgG |
| 6K5 | |
| Monoclonal | |
| Rabbit | |
| 5195 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.