Learn More
Tocris Bioscience™ PF 5274857 hydrochloride
Description
PF 5274857 hydrochloride is a high affinity and selective Smoothened (Smo) receptor antagonist (Ki = 4.6 nM). Exhibits >1000-fold selectivity for Smo over a panel of other receptors, ion channels and enzymes. Inhibits Shh-induced Hedgehog signaling in MEF cells. Induces tumor regression in a medulloblastoma allograft mouse model. Orally available and brain penetrant.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | PF 5274857 hydrochloride |
| Purity | >98% |
| Molecular Weight (g/mol) | 473.42 |
| Inhibitors | PF 5274857 hydrochloride |