missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PF 5274857 hydrochloride
GREENER_CHOICE

Product Code. 18795578
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18795578 10 mg 10mg
18734729 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18795578 Supplier Tocris Bioscience™ Supplier No. 5262/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity and selective Smoothened (Smo) receptor antagonist

PF 5274857 hydrochloride is a high affinity and selective Smoothened (Smo) receptor antagonist (Ki = 4.6 nM). Exhibits >1000-fold selectivity for Smo over a panel of other receptors, ion channels and enzymes. Inhibits Shh-induced Hedgehog signaling in MEF cells. Induces tumor regression in a medulloblastoma allograft mouse model. Orally available and brain penetrant.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type PF 5274857 hydrochloride
Purity >98%
Molecular Weight (g/mol) 473.42
Inhibitors PF 5274857 hydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.