Learn More
Savillex™ PFA Digestion Vessels
Product Code. 11912195
Change view
Click to view available options
Capacity (Metric):
60 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11912195 60 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11912195 Supplier Savillex™ Supplier No. 30006003
  • ONLINE EXCLUSIVE PRICE
    178.92€
    6.13€ / Pack of 1
    Save 172.79€
    96% Off
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Product Type Digestion Vessel
Capacity (Metric) 60 mL
For Use With (Application) Rapid sample digestion via microwave techniques
Material PFA