missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer PFA Tubing
Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1/16 in.
1/2 in.
1/8 in.
3/16 in.
3/32 in.
5/32 in.
7/16 in.
Diameter (English) Outer:
1/2 in.
1/4 in.
1/8 in.
3/16 in.
5/32 in.
9/16 in.
Packungsgröße:
15m
Beschreibung
Made from 100% virgin ultra-high molecular weight perfluoroalko in., y. E in., cellent fle in., ibility compared to other PFA grade tubing. Wide temperature range of -160 to 260 °C (-320 to 500 °F). Does not contain BPA, phthalates, or PVC. Sterilize by autoclave, EtO, and chemical sterilants. Complies with FDA 21 CFR 177.1550, USP Class VI, ASTM D 6867-03. Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Spezifikation
Spezifikation
| Certifications/Compliance | ADCF;FDA;Non-Phthalate;RoHS;USP |
| Diameter (English) Inner | 5/32 in. |
| Diameter (English) Outer | 1/4 in. |
| Overall Length (English) | 50 ft. |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur