missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PFI 653

Codice prodotto. 18774444
Cambia vista
Click to view available options
Quantity:
10 mg
50 mg
Dimensione della confezione:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
18774444 10 mg 10mg
18744524 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18774444 Fornitore Tocris Bioscience™ N. del fornitore 7963/10

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Potent and selective vanin 1 inhibitor

PFI 653 is a potent and selective vanin 1 inhibitor (IC50 values in the range 6.85 - 24.5 nM). The compound exhibits selectivity over a panel of 40 kinases and a selection of 35 other targets including enzymes, receptors and ion channels (IC50 vs biotinidase >50 μM). Following administration of a single dose of PFI 653 (2 mg/kg iv) in rats AUC is 1790 ng.h/mL, half-life is 1 h and clearance is 19 mL/min/kg. Following oral administration of PFI 653 in solution, bioavailablility is 96%. In mice with DSS-induced inflammatory bowel disease (IBD), oral adminstration of PFI 653 dose-dependently reduces vanin 1 enzyme activity and disease symptoms.
TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material 8-Oxa-2-azaspiro[4.5]dec-2-yl[2-[(2-pyrazinylmethyl)amino]-5-pyrimidinyl]methanone
CAS 2173134-00-2
Quantity 10 mg
Target Vanin-1/VNN1
Molecular Formula C18H22N6O2
Purity 0.98
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.