Learn More
PGBD3 Monoclonal Antibody (OTI4G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810380S
Description
PGBD3 Monoclonal antibody specifically detects PGBD3 in Human samples. It is validated for Western BlotSpecifications
| PGBD3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NR83 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 56731 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4G9 | |
| Unconjugated | |
| SLC2A4RG | |
| 4732403I04; ATP-dependent helicase ERCC6; C130058G22Rik; Cockayne syndrome protein CSB; CS group B correcting; Csb; DNA excision repair protein ERCC-6; Ercc6; excision repair cross-complementation group 6; excision repair cross-complementing rodent repair deficiency, complementation group 6; FLJ90201; PGBD3; piggyBac transposable element derived 3; piggyBac transposable element-derived protein 3 | |
| Human recombinant protein fragment corresponding to amino acids 1-259 of human SLC2A4RG produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.