missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGM1 Antibody (0F6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43016
This item is not returnable.
View return policy
Description
PGM1 Monoclonal antibody specifically detects PGM1 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| PGM1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 0F6 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/ Immunofluorescence 1:20 - 1:200 | |
| EC 5.4.2, EC 5.4.2.2, Glucose phosphomutase 1, GSD14, PGM 1, phosphoglucomutase 1, phosphoglucomutase-1 | |
| phosphoglucomutase 1 - (Uniprot# P36871) | |
| 100 μg | |
| Lipid and Metabolism | |
| 5236 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction