Learn More
PGM1 Antibody (CL3299) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61618CL1
Product Code. 30298704
  • 1175.00€ / 0.10mL
Description
PGM1 Monoclonal antibody specifically detects PGM1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PGM1
Monoclonal
CoraFluor 1
EC 5.4.2, EC 5.4.2.2, Glucose phosphomutase 1, GSD14, PGM 1, phosphoglucomutase 1, phosphoglucomutase-1
This antibody was developed using a recombinant protein derived from P36871, with the exact immunogen sequence remaining proprietary.
0.1 mL
Lipid and Metabolism
5236
Store at 4⪚C in the dark. Do not freeze.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL3299
PBS
Mouse
Protein A purified