Learn More
PGM2L1 Monoclonal Antibody (OTI2D12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812135S
Description
PGM2L1 Monoclonal antibody specifically detects PGM2L1 in Human, Rat samples. It is validated for Western BlotSpecifications
| PGM2L1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P40939, Q64428 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 170670, 3030 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2D12 | |
| Unconjugated | |
| HADHA | |
| 4931406N15Rik; AI553438; BM32A; FLJ32029; Glucose 1,6-bisphosphate synthase; im:7140576; im:7140576 protein; Pgm2l1; phosphoglucomutase 2 like 1; phosphoglucomutase 2-like 1; phosphoglucomutase-2-like 1; PMMLP; zgc:198285 | |
| Human recombinant protein fragment corresponding to amino acids 106-325 of human HADHA produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human, Rat | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.