missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17149-1-AP-20UL
This item is not returnable.
View return policy
Description
PGS1 functions in the biosynthesis of the anionic phospholipids phosphatidylglycerol and cardiolipin.Specifications
| PGS1 | |
| Polyclonal | |
| Unconjugated | |
| PGS1 | |
| PGP synthase 1, PGS1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 303698, 74451, 9489 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q32NB8, Q8BHF7 | |
| Pgs1 | |
| PGS1 Fusion Protein Ag10961 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction