Learn More
Consort™ C3050T Bio-Electronic Analyzer Kit
Product Code. 11742029
Change view
Click to view available options
Range (mV):
+2000 mV
pH Range:
-2 to 16
Temperature (Metric) Range:
-5°C to 105°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range (mV) pH Range Temperature (Metric) Range Unit Size
11742029 +2000 mV -2 to 16 -5°C to 105°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11742029 Supplier Consort™ Supplier No. C3050T
  • ONLINE EXCLUSIVE PRICE
    3647.63€
    2518.00€ / Pack of 1
    Save 1129.63€
    30% Off
In Stock
Añadir a la cesta
Edge
Añadir a la cesta
missing translation for 'validMainframeMsg'
A bio-electronic analyser kit capable of measuring all parameters according to Vincent's method, supplied complete with meter, pH and conductivity electrodes, buffers, conductivity standards, and electrode holders.
  • Supplied with meter and pH/ORP electrode SP35B, conductivity electrode SK20T, 2 x 50mL buffers (pH 4 and 7), 50mL conductivity standard (0.01 M KCI), 50mL electrolyte (3M KCI), 50mL redox standard (358 mV), flexible electrode holder SH300
  • All 3 gold plated BNC connectors each accept different electrodes of which 2 connectors also accept conductivity electrodes
  • Multichannel for up to three simultaneous measurements
  • Each channel can measure 2 times per second
  • No interference between pH/ORP and conductivity electrodes in the same solution
  • Custom calibration tables allow the user to add complete buffer/standard-temperature relation tables
  • Tables can be entered via a device menu or uploaded from a PC
  • Direct rH&sub2; and μW measurement when using a pH, ORP and EC electrode
  • Wide display shows all channels at the same time including temperature and date/time
  • Stability indicator ensures visualization when the measurement has stabilized
  • Stability algorithm ensures stable readings with ability to detect fast changes
  • Hold function allows the display to be frozen
  • Selectable resolution for more stable readings on mV and pH
  • Range lock for conductivity measurements
  • Capacitive compensation eliminates the capacitive component of the electrode and cable when measuring low conductivities
  • Galvanic isolated USB interface eliminates ground loop effects when connected to a PC
  • Pre-programmed standards for pH: 1.68, 2.00, 4.00, 4.01, 6.87, 6.99, 9.18, 9.21, 10.01, 12.00, 12.45 (at 25°C)
  • Pre-programmed standards for conductivity: 1413 μS/cm, 12.88 mS/cm, 111.8 mS/cm (at 25°C)
  • Free software and firmware updates downloadable from www.consort.be
Especificaciones
Accuracy (pH) 0.01% ±1 digit
Accuracy (mV) 0.01% ±1 digit
Calibration Points (pH) 1 to 5
Type Multiparameter Analyzer Kit
Includes 3050T Meter, Electrode (pH/ORP), Conductivity Probe, pH Buffer and Conductivity Standards, Electrolyte Solution and ORP Electrode Holder Arm
Model C3050T
pH Range -2 to 16
Range (mV) +2000 mV
Relative Humidity Range 0 to 42 rh
Resolution (mV) 0.1 mV
Resolution (pH) 0.001 pH
Temperature (Metric) Range -5°C to 105°C
Mostrar más Mostrar menos
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.