Product Code. 11758452
Shop All Fisherbrand Products
Click to view available options
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Connector Types
Unit Size
11758452
S7
Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code.
11758452
153.96€ 96.99€ / Pack of 1 Save 56.97€ 37% Off
Estimated Shipment: 09-10-2026Log in to see stock.
Regulatory: This product is under eco-contribution tax.
This uniquely-designed ORP pH electrode features a S7 head epoxy body that is ideal for many laboratory and other scientific environments.
ORP Electrode redox head S7 Epoxy body 12mm x 120mm
Specifications
Connector Types
S7
Length (Metric)
120 mm
Diameter (Metric)
12 mm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?