Learn More
Hanna Instruments™ HI-122-02 Benchtop pH/ORP/Temperature Meter with Printer
Product Code. 11785645
Change view
Click to view available options
Range (pH):
-2 to 16.00
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range (pH) unitSize
11785645 -2 to 16.00 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11785645 Supplier Hanna Instruments™ Supplier No. HI12202
Only null left
Add to basket
Add to basket
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
This meter offers accurate pH measurement with the added feature of a built-in printer. With millesimal resolution and automatic temperature compensation, HI-122-02 is provided with the Calibration Check™ system that evaluates electrode condition and function in relation to the calibration.
  • Buil-in printer
  • Calibration Check™ system
  • 5-point calibration
  • Adheres to GLP
  • Diameter: 203 mm (8 in.)
Specifications
Accuracy (ORP) ±0.2 mV (± 699.9 mV) /±0.5 mV (± 999.9 mV) /±1 mV (±2000 mV)
Accuracy (pH) ±0.01/±0.002
Calibration Automatic, Up to 5 points with 7 memorized values and 2 custom buffers
Calibration Points (pH) 1.68, 4.01, 6.86, 7.01, 9.18, 10.01, 12.45
Detection Range -2°C to +16.00°C
Display Type LCD
Range (mV) ±999.9 mV; ±2000 mV
Range (pH) -2 to 16.00
Resolution (pH) 0.01/0.001
Temperature Range (Metric) -20°C to +120°C
Includes 1 m (3.3 ft.) Cable
Inputs BNC
Model HI12202
Power Supply 12 VDC
Product Type pH/ORP/Temperature Meter
Range (ORP) ±999.9/±2000 mV
Range (Relative mV) ±2000 mV
Resolution (mV) 0.1/1 mV
Resolution (ORP) 0.1/1 mV
Temperature Accuracy ±0.4°C
Temperature Compensation Automatic or Manual, -20.0°C to +120°C
Temperature Probe Calibration HI 7662-T
Temperature Resolution 0.1°C
Width (English) 11 in.
Width (Metric) 280 mm
Height (English) 3.3 in.
Height (Metric) 84 mm
Weight (Metric) 1.8 kg
Voltage 230 V
Frequency 50 Hz