Learn More
Christeyns™ Phago'Wipes Spores Wipes
Product Code. 17156464
Change view
Click to view available options
Packaging Quantity:
100 Wipes/Pk., 6 Pk./Cs.
Unit Size:
Pack of 600
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Unit Size
17156464 100 Wipes/Pk., 6 Pk./Cs. Pack of 600
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17156464 Supplier Christeyns™ Supplier No. 60012
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Phago'Wipes Spores Wipes are bactericidal, fungicidal, mycobactericidal, virucidal and sporicidal with peracetic acid as their active ingredient.
  • Disinfectant detergent wipes for all surfaces, including non-invasive medical devices (operating table, operating light, monitors, cables and connectors, stethoscopes, reflex hammers, blood pressure monitors)
  • Can be used, with rinsing, on surfaces that may be in direct contact with foodstuffs
  • Formulated from an active substance (peracetic acid) guaranteeing broad-spectrum microbiological efficacy while ensuring respect for the environment through its biodegradability into acetic acid, oxygen and water
  • Ready-to-use
Specifications
Product Type Sanitizing Wipes
Length (English) 6.7 in.
Length (Metric) 17 cm
Width (English) 4.7 in.
Width (Metric) 12 cm
Material Plastic
Packaging Quantity 100 Wipes/Pk., 6 Pk./Cs.
compliance-icons
Product Identifier
  • PHAGO'SPORE
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: hydrogen peroxide, propan-2-ol