missing translation for 'onlineSavingsMsg'
Learn More

Phantolide, TRC

Product Code. 30135622
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135622 500 mg 500mg
17951145 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135622 Supplier Toronto Research Chemicals Supplier No. TRCP294650500MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15323-35-0
Molecular Formula C17 H24 O
Molecular Weight (g/mol) 244.37
Synonym Ketone, 1,1,2,3,3,6-hexamethyl-5-indanyl methyl (6CI,8CI), 1-(2,3-Dihydro-1,1,2,3,3,6-hexamethyl-1H-inden-5-yl)ethanone, 1-(1,1,2,3,3,6-Hexamethyl-2,3-dihydro-1H-inden-5-yl)ethan-1-one, 6-Acetyl-1,1,2,3,3,5-hexamethylindan, 6-Acetyl-1,1,2,3,3,5-hexamethylindane, AHDI, AHMI, Musk phantolid, NSC 16827, Phantolid, Phantolide, Ethanone, 1-(2,3-dihydro-1,1,2,3,3,6-hexamethyl-1H-inden-5-yl)-
IUPAC Name 1-(1,1,2,3,3,6-hexamethyl-2H-inden-5-yl)ethanone
SMILES CC1C(C)(C)c2cc(C)c(cc2C1(C)C)C(=O)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C17H24O/c1-10-8-14-15(9-13(10)11(2)18)17(6,7)12(3)16(14,4)5/h8-9,12H,1-7H3
Formula Weight 244.1827
Chemical Name or Material Phantolide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.