Learn More
Kirsch Philipp™ Pharmaceutical Refrigerator
Product Code. 10630615
Change view
Click to view available options
Capacity (Metric) Freezer:
460 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Freezer Unit Size
10630615 460 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10630615 Supplier Kirsch Philipp™ Supplier No. 10589
  • ONLINE EXCLUSIVE PRICE
    6351.45€
    5709.00€ / Pack of 1
    Save 642.45€
    10% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes
Ten height adjustable drawers
Storage for sensitive chilled goods.
  • Individual drawer division with adjustable length and cross dividers
  • Recirculating air-cooling
  • Optimized ventilation plates to minimize interior temperature differences
  • Automatic defrosting
  • Melt water evaporation
  • Easy-to-read LED temperature display
  • Easy accessible control panel with key switch
  • Micro-processor controlled temperature
  • Thawing regulation to maximize temperature stability
  • Minimum/Maximum temperature memory
  • Safety control to prevent freezing
  • Optical and acoustical alarm including power failure alarm
  • Potential free contact for alarm transmission
  • RS-485 interface
  • Low-noise compressor
Specifications
Capacity (Metric) Freezer 460 L
Temperature Range (Refrigerator) 2°C to 20°C
Capacity (Metric) Shelf 24 kg
For Use With (Application) Storage of pharmaceutical products
Dimensions (D x W x H) Interior 57 x 60 x 128 cm
Energy Usage 140 kw/24 hours
Voltage 220/240 V
Electrical Requirements 50 - 60 Hz 180 w
Weight (Metric) 143 kg (gross)