missing translation for 'onlineSavingsMsg'
Learn More

Phenindione, TRC

Product Code. 30426892
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426892 25 g -
30423994 1 g 1g
30420052 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30426892 Supplier Toronto Research Chemicals Supplier No. TRCP29725025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-12-5
Molecular Formula C15 H10 O2
Molecular Weight (g/mol) 222.24
Synonym 1H-Indene-1,3(2H)-dione, 2-phenyl-, 1,3-Indandione, 2-phenyl- (6CI,8CI), 2-Phenyl-1,3-diketohydrindene, 2-Phenyl-1,3-indandione, 2-Phenyl-1,3-indanedione, 2-Phenyl-1H-indene-1,3(2H)-dione, 2-Phenylindandione, Athrombon, Bindan, Cronodione, Danedion, Danilon, Danilone, Diadilan, Dindevan, Dineval, Diophindane, Emandion, Emandione, Fenhydren, Fenilin, Fenindion, Hedulin, Hemolidione, Indema, Indion, Indon, NSC 41693, PID, Phenhydren, Phenillin, Phenindione, Phenylen, Phenylin, Phenylindione, Pindione, Rectadione, Theradione, Thrombasal, Tromazal, Trombol
IUPAC Name 2-phenylindene-1,3-dione
SMILES O=C1C(C(=O)c2ccccc12)c3ccccc3

Specifications

Quantity 25 g
InChI Formula InChI=1S/C15H10O2/c16-14-11-8-4-5-9-12(11)15(17)13(14)10-6-2-1-3-7-10/h1-9,13H
Formula Weight 222.0681
Chemical Name or Material Phenindione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.