missing translation for 'onlineSavingsMsg'
Learn More

Phenmedipham, TRC

Product Code. 30135332
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135332 5 g 5g
17993745 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135332 Supplier Toronto Research Chemicals Supplier No. TRCP3146005G
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13684-63-4
Molecular Formula C16 H16 N2 O4
Molecular Weight (g/mol) 300.31
Synonym Carbamic acid, N-(3-methylphenyl)-, 3-[(methoxycarbonyl)amino]phenyl ester, Carbamic acid, (3-methylphenyl)-, 3-[(methoxycarbonyl)amino]phenyl ester (9CI), Carbanilic acid, m-hydroxy-, methyl ester, m-methylcarbanilate (ester) (8CI), Carbanilic acid, m-methyl-, ester with methyl m-hydroxycarbanilate (8CI), 3-(Carbomethoxyamino)phenyl 3-methylcarbanilate, 3-[(Methoxycarbonyl)amino]phenyl N-(3-methylphenyl)carbamate, Betanal, Fenifan, Fenmedifam, Kemifam, Kemifam FL, Kontakt, Kontakt (herbicide), Kontakt FCS, Methyl 3-(m-tolylcarbamoyl)carbanilate, Methyl N-[3-[N-(3-methylphenyl)carbamoyloxy]phenyl]carbamate, Methyl N-[3-[N'-(3'-methylphenyl)carbamoyloxy]phenyl]carbamate, Methyl m-hydroxycarbanilate m-methylcarbanilate, Phenmedipham, SN 38584, Stepham, Synbetan P, Vangard
IUPAC Name [3-(methoxycarbonylamino)phenyl] N-(3-methylphenyl)carbamate
SMILES COC(=O)Nc1cccc(OC(=O)Nc2cccc(C)c2)c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C16H16N2O4/c1-11-5-3-6-12(9-11)18-16(20)22-14-8-4-7-13(10-14)17-15(19)21-2/h3-10H,1-2H3,(H,17,19)(H,18,20)
Formula Weight 300.111
Chemical Name or Material Phenmedipham
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.