missing translation for 'onlineSavingsMsg'
Learn More

Phenmetetrazine Hydrochloride, TRC

Product Code. 30419171
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30419171 50 mg 50mg
30412152 25 mg 25mg
30413406 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30419171 Supplier Toronto Research Chemicals Supplier No. TRCP29640050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenmetetrazine Hydrochloride
Molecular Formula C13 H19 N O . H Cl
Quantity 50 mg
Synonym 4-Ethyl-3-methyl-2-phenyl-morpholine Hydrochloride
InChI Formula InChI=1S/C13H19NO.ClH/c1-3-14-9-10-15-13(11(14)2)12-7-5-4-6-8-12;/h4-8,11,13H,3,9-10H2,1-2H3;1H
SMILES Cl.CCN1CCOC(C1C)c2ccccc2
IUPAC Name 4-ethyl-3-methyl-2-phenylmorpholine;hydrochloride
Molecular Weight (g/mol) 241.757
Formula Weight 241.123
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.