missing translation for 'onlineSavingsMsg'
Learn More

Phenol, Fused, U.S.P., J.T. Baker™

Product Code. 15508064 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2 kg
Packaging:
Narrow Mouth PVC Coated Amber Bottle
Unit Size:
2kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15508064 2 kg Narrow Mouth PVC Coated Amber Bottle 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15508064 Supplier Avantor J.T.Baker Supplier No. 286205

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Spécification

CAS 108-95-2
Flash Point 79.00°C
Packaging Narrow Mouth PVC Coated Amber Bottle
Molecular Formula C6H6O
MDL Number MFCD00002143
Quantity 2 kg
Specific Gravity 1L = 1.06kg
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILES OC1=CC=CC=C1
IUPAC Name phenol
Molecular Weight (g/mol) 94.11
PubChem CID 996
ChEBI CHEBI:15882
Formula Weight 94.11
Grade USP
Chemical Name or Material Phenol, Fused
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.