missing translation for 'onlineSavingsMsg'
Learn More

Phenol, TRC

Product Code. 30041165
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041165 5 g 5g
30011547 100 g 100g
30005244 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041165 Supplier Toronto Research Chemicals Supplier No. TRCP3180055G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6 H6 O
Molecular Weight (g/mol) 94.11
Synonym Phenol, Guaiacol Imp. B (EP), Hexylresorcinol Imp. A (EP), Salicylic Acid Imp. C (EP), 2-Allphenol, Benzenol, Carbolic acid, ENT 1814, Hydroxybenzene, Monohydroxybenzene, Monophenol, NSC 36808, Oxybenzene, Phenic acid, Phenyl alcohol, Phenyl hydrate, Phenyl hydroxide, Phenylic acid, Phenylic alcohol
IUPAC Name phenol
SMILES Oc1ccccc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H
Formula Weight 94.0419
Percent Purity >95%
Chemical Name or Material Phenol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.