missing translation for 'onlineSavingsMsg'
Learn More

Phenothiazine, 98+%

Product Code. p-7020529
Change view
Click to view available options
Quantity:
250 g
500 g
5000 g
Unit Size:
250g
5000g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494164 250 g 250g
11498680 500 g 500g
11404174 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494164 Supplier Thermo Scientific Alfa Aesar Supplier No. A12517.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 92-84-2 | C12H9NS | 199.271 g/mol

It acts on type I calcium/calmodulin dependent phosphodiesterase and also a useful electron donor. Phenothiazine finds uses in metal free organic dye sensitizers, dyes and antioxidants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It acts on type I calcium/calmodulin dependent phosphodiesterase and also a useful electron donor. Phenothiazine finds uses in metal free organic dye sensitizers, dyes and antioxidants.

Solubility
Soluble in water (0.13 mg/ml at 23°C), ethanol, acetone, and ether. Insoluble in chloroform.

Notes
Store under dry inert gas. Protect from sunlight. Ensure good ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-84-2
Molecular Formula C12H9NS
Molecular Weight (g/mol) 199.271
MDL Number MFCD00005015
InChI Key WJFKNYWRSNBZNX-UHFFFAOYSA-N
Synonym phenothiazine, thiodiphenylamine, feeno, dibenzothiazine, phenosan, phenthiazine, dibenzo-1,4-thiazine, penthazine, souframine, agrazine
PubChem CID 7108
ChEBI CHEBI:37931
IUPAC Name 10H-phenothiazine
SMILES C1=CC=C2C(=C1)NC3=CC=CC=C3S2

Specifications

Melting Point 182°C to 186°C
Boiling Point 235°C (21 mmHg)
Quantity 250 g
Beilstein 143237
Sensitivity Light sensitive
Merck Index 14,7252
Solubility Information Soluble in water (0.13mg/ml at 23°C),ethanol,acetone,and ether. Insoluble in chloroform.
Formula Weight 199.28
Percent Purity ≥98%
Chemical Name or Material Phenothiazine
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenothiazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Phenothiazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.