Learn More
Phenyl 1-Thio-Beta-D-galactoside, TRC
Specifications
Specifications
| Chemical Name or Material | Phenyl 1-Thio-Beta-D-galactoside |
| CAS | 16758-34-2 |
| Molecular Formula | C12 H16 O5 S |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C12H16O5S/c13-6-8-9(14)10(15)11(16)12(17-8)18-7-4-2-1-3-5-7/h1-5,8-16H,6H2/t8-,9+,10+,11-,12+/m1/s1 |
| SMILES | OC[C@H]1O[C@@H](Sc2ccccc2)[C@H](O)[C@@H](O)[C@H]1O |
| IUPAC Name | (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-phenylsulfanyloxane-3,4,5-triol |
| Molecular Weight (g/mol) | 272.32 |
| Formula Weight | 272.0718 |