missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Dichlorophosphate, TRC

Product Code. 30410993
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410993 100 g 100g
30417712 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30410993 Supplier Toronto Research Chemicals Supplier No. TRCP319950100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenyl Dichlorophosphate
CAS 770-12-7
Molecular Formula C6H5Cl2O2P
Quantity 100 g
Synonym Phosphorodichloridic Acid Phenyl Ester, Phenyl Phosphorodichloridate, Dichlorophenoxyphosphine Oxide, NSC 44412, Phenoxydichlorophosphine Oxide, Phenoxyphosphoryl Dichloride, Phenyl Dichlorophosphinate, Phenylphosphoric Dichloride
InChI Formula InChI=1S/C6H5Cl2O2P/c7-11(8,9)10-6-4-2-1-3-5-6/h1-5H
SMILES ClP(=O)(Cl)Oc1ccccc1
IUPAC Name dichlorophosphoryloxybenzene
Molecular Weight (g/mol) 210.98
Formula Weight 209.9404
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.