missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Phenyl methacrylate, 95%, stab.

Product Code. 11358535
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358535 25 g 25g
11348535 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358535 Supplier Alfa Aesar™ Supplier No. L11978.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
Phenyl methacrylate is used as thermoset acrylic resin, fiber finishing agent and comonomers of resin lens.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2177-70-0
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00048117
InChI Key QIWKUEJZZCOPFV-UHFFFAOYSA-N
Synonym phenyl methacrylate, phenylmethacrylate, methacrylic acid, phenyl ester, 2-propenoic acid, 2-methyl-, phenyl ester, acmc-20ajx1, phenyl 2-methylacrylate #, methacrylic acid phenyl ester, phenyl methacrylate stabilized with bht
PubChem CID 75121
IUPAC Name phenyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC1=CC=CC=C1

Specifications

Density 1.052
Boiling Point 115°C to 116°C (10 mmHg)
Flash Point 60°C (140°F)
Refractive Index 1.515
Quantity 25 g
UN Number UN3272
Beilstein 1907503
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 162.19
Percent Purity 95%
Chemical Name or Material Phenyl methacrylate, stabilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.