missing translation for 'onlineSavingsMsg'
Learn More

Phenyldimethylchlorosilane, TRC

Product Code. 30409607
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30409607 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409607 Supplier Toronto Research Chemicals Supplier No. TRCP3197351G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenyldimethylchlorosilane
CAS 768-33-2
Molecular Formula C8H11ClSi
Quantity 1 g
Synonym (Chlorodimethylsilyl)benzene, Chlorodimethylphenylsilane, Dimethylphenylchlorosilane, Phenyldimethylsilyl Chloride
InChI Formula InChI=1S/C8H11ClSi/c1-10(2,9)8-6-4-3-5-7-8/h3-7H,1-2H3
SMILES C[Si](C)(Cl)c1ccccc1
IUPAC Name chloro-dimethyl-phenylsilane
Molecular Weight (g/mol) 170.71
Formula Weight 170.0319
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.