missing translation for 'onlineSavingsMsg'
Learn More

Phenylephrine EP Impurity E (as Hydrochloride), Mikromol™

Product Code. 17950626
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17950626 100 mg 100mg
17922256 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17950626 Supplier Mikromol Supplier No. MM0081.08
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Phenylephrine EP Impurity E (as Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Benzyl-methylamino)-1-(3-hydroxyphenyl)ethanone Hydrochloride (Benzylphenylephrone Hydrochloride)
CAS 71786-67-9
Analyte or Component Names 2-(Benzyl-methylamino)-1-(3-hydroxyphenyl)ethanone Hydrochloride (Benzylphenylephrone Hydrochloride)
Shipping Condition Room Temperature
Molecular Formula C16 H17 N O2 . Cl H
Certifications/Compliance ISO 17025
Synonym Phenylephrine Hydrochloride Imp. E (EP), Phenylephrine Imp. E (EP), Phenylephrine USP Related Compound E, Phenylephrine USP RC E, 2-(Benzyl-methylamino)-1-(3-hydroxyphenyl)ethanone Hydrochloride, Phenylephrine Hydrochloride Imp. E (EP) as Hydrochloride, Benzylphenylephrone Hydrochloride, Phenylephrine Related Compound E as Hydrochloride, Phenylephrine Hydrochloride Impurity E as Hydrochloride, Phenylephrine Impurity E as Hydrochloride, Phenylephrine Impurity E as Hydrochloride
InChI Formula InChI=1S/C16H17NO2.ClH/c1-17(11-13-6-3-2-4-7-13)12-16(19)14-8-5-9-15(18)10-14;/h2-10,18H,11-12H2,1H3;1H
SMILES Cl.CN(CC(=O)c1cccc(O)c1)Cc2ccccc2
IUPAC Name 2-[benzyl(methyl)amino]-1-(3-hydroxyphenyl)ethanone;hydrochloride
Molecular Weight (g/mol) 291.77
Quantity 100 mg
Formula Weight 291.1026 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.