missing translation for 'onlineSavingsMsg'
Learn More

Phenylethylacetylurea (2-Phenylbutyrylurea), Mikromol™

Product Code. 17960576
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17960576 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17960576 Supplier Mikromol Supplier No. MM0265.01
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Phenylethylacetylurea (2-Phenylbutyrylurea)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Phenylbutyrylurea
CAS 90-49-3
Analyte or Component Names 2-Phenylbutyrylurea
Shipping Condition Room Temperature
Molecular Formula C11 H14 N2 O2
Certifications/Compliance ISO 17034
Synonym 2-Phenylbutyrylurea, Urea, (2-phenylbutyryl)- (6CI,7CI,8CI), 1-[(Ethyl)phenylacetyl]urea, Benuride, EPA, Ethylphenacemide, Lircapyl, M 551, PBU, Pheneturide, Phenuride, Phenylethylacetylurea, S 46, dl-Pheneturide, Benzeneacetamide, N-(aminocarbonyl)-alpha-ethyl-, N-(Aminocarbonyl)-alpha-ethylbenzeneacetamide, N-(alpha-Phenylbutyryl)urea, alpha-Phenyl-alpha-ethylacetylurea
InChI Formula InChI=1S/C11H14N2O2/c1-2-9(10(14)13-11(12)15)8-6-4-3-5-7-8/h3-7,9H,2H2,1H3,(H3,12,13,14,15)
SMILES CCC(C(=O)NC(=O)N)c1ccccc1
IUPAC Name N-carbamoyl-2-phenylbutanamide
Molecular Weight (g/mol) 206.24
Quantity 100 mg
Formula Weight 206.1055 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.