Learn More
Phenylhydrazine, TRC
Product Code. 30011916
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011916 500 g 500g
30003681 100 g 100g
18041420 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011916 Supplier Toronto Research Chemicals Supplier No. TRCP335075500G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 100-63-0
Molecular Formula C6 H8 N2
Molecular Weight (g/mol) 108.14
Synonym Hydrazine, phenyl-, Phenylhydrazine, Hydrazinobenzene, Monophenylhydrazine
IUPAC Name phenylhydrazine
SMILES NNc1ccccc1
Specifications
Quantity 500 g
InChI Formula InChI=1S/C6H8N2/c7-8-6-4-2-1-3-5-6/h1-5,8H,7H2
Formula Weight 108.0687
Chemical Name or Material Phenylhydrazine