missing translation for 'onlineSavingsMsg'
Learn More

Phenylhydroquinone diacetate, 98%, Thermo Scientific™

Product Code. 11409417
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11409417 10 g 10g
11419417 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11409417 Supplier Thermo Scientific Alfa Aesar Supplier No. B21433.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58244-28-3
Molecular Formula C16H14O4
Molecular Weight (g/mol) 270.28
MDL Number MFCD00145106
InChI Key DZVDHXPXHBVBNZ-UHFFFAOYSA-N
Synonym phenylhydroquinone diacetate, 2,5-diacetoxybiphenyl, biphenyl-2,5-diyl diacetate, 1,1'-biphenyl-2,5-diol diacetate, 1,1'-biphenyl-2,5-diyl diacetate, 1,1'-biphenyl-2,5-diol, diacetate, biphenyl-2,5-diol diacetate, phenylhydrochinondiacetat, phenyl hydroquinone diacetate
PubChem CID 94584
IUPAC Name (4-acetyloxy-3-phenylphenyl) acetate
SMILES CC(=O)OC1=CC=C(OC(C)=O)C(=C1)C1=CC=CC=C1

Specifications

Melting Point 67°C to 70°C
Quantity 10 g
Beilstein 2383464
Formula Weight 270.29
Percent Purity 98%
Chemical Name or Material Phenylhydroquinone diacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.