Learn More
PHF21B Monoclonal Antibody (OTI5H9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503284S
Description
PHF21B Monoclonal antibody specifically detects PHF21B in Human samples. It is validated for Western BlotSpecifications
| PHF21B | |
| Monoclonal | |
| 0.41 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NXA8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23408 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5H9 | |
| Unconjugated | |
| SIRT5 | |
| A730032D07Rik; BHC80L; KIAA1661; PHD finger protein 21B; PHD finger protein 4; PHF21B; PHF4; RGD1308739 | |
| Full length human recombinant protein of human SIRT5 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.